Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
TC06000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
ENST00000375651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
ENST00000375652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
ENST00000458062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
ENST00000541556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003955.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001271.hg.1 |
TC6_cox_hap2000078.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001271.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
TC06000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
ENST00000375651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
ENST00000375652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
ENST00000458062.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
ENST00000541556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003954.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001226.hg.1 |
TC6_qbl_hap6000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001226.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000500.hg.1 |
TC6_apd_hap1000039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000500.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001214.hg.1 |
TC6_dbb_hap3000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001214.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003956.hg.1 |
TC06000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003956.hg.1 |
ENST00000375651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003956.hg.1 |
ENST00000375652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003956.hg.1 |
ENST00000541556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003956.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152129.hg.1 |
TC0600007613.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000375651.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
ENST00000608703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007613.hg.1 |
NM_005345.5 |
Refseq |