Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039600.hg.1 |
TC01002545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039600.hg.1 |
ENST00000372844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039600.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039598.hg.1 |
TC01002545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039598.hg.1 |
ENST00000372844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039598.hg.1 |
ENST00000450300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039598.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039595.hg.1 |
TC01002545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039595.hg.1 |
ENST00000372844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039595.hg.1 |
ENST00000450300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039595.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039596.hg.1 |
TC01002545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039596.hg.1 |
ENST00000372844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039596.hg.1 |
ENST00000450300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039596.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039594.hg.1 |
TC01002545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039594.hg.1 |
ENST00000372844.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039594.hg.1 |
ENST00000450300.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039594.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203219.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013802.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203216.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100203218.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203218.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203218.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203218.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203218.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000612703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_001282397.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203220.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
ENST00000617690.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
NM_001282396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203215.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100203221.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203221.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203221.hg.1 |
ENST00000372844.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100203221.hg.1 |
NM_016257.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100203221.hg.1 |
TC0100013802.hg.1 |
Clariom D TC |