Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002245.hg.1 |
TC0X000200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002245.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002245.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002245.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002245.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002240.hg.1 |
TC0X000200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002240.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002240.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002240.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002241.hg.1 |
TC0X000200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002241.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002241.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002243.hg.1 |
TC0X000200.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002243.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002243.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002243.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147906.hg.1 |
TC0X00007034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147908.hg.1 |
TC0X00007034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147902.hg.1 |
TC0X00007034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147903.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147903.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147903.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147903.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147903.hg.1 |
TC0X00007034.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147905.hg.1 |
TC0X00007034.hg.1 |
Clariom D TC |