Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009412.hg.1 |
ENST00000426627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
ENST00000373644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007861.hg.1 |
NM_030625.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009412.hg.1 |
ENST00000426627.1 |
Ensembl |