Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007392.hg.1 |
ENST00000554138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007392.hg.1 |
ENST00000554138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004182.hg.1 |
ENST00000430451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004182.hg.1 |
ENST00000446982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004182.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004182.hg.1 |
TC14000375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004180.hg.1 |
ENST00000430451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004180.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004180.hg.1 |
TC14000375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004181.hg.1 |
ENST00000430451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004181.hg.1 |
ENST00000446982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004181.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004181.hg.1 |
TC14000375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004183.hg.1 |
ENST00000430451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004183.hg.1 |
ENST00000446982.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004183.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004183.hg.1 |
TC14000375.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151551.hg.1 |
TC1400007395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151549.hg.1 |
TC1400007395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151550.hg.1 |
TC1400007395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151547.hg.1 |
TC1400007395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
ENST00000568344.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151548.hg.1 |
TC1400007395.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007392.hg.1 |
ENST00000554138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007392.hg.1 |
ENST00000554138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007392.hg.1 |
ENST00000554436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000430451.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000555409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
ENST00000568344.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007395.hg.1 |
NM_031914.2 |
Refseq |