Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
TC10002963.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
ENST00000358140.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
ENST00000359178.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
ENST00000414655.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
ENST00000545298.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
NM_001040084.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015454.hg.1 |
NM_001098845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000577813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000583874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000583911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000585281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000611843.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001040084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001271702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001271703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000577813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000583874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000583911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000585281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
ENST00000611843.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001040084.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001271702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
NM_001271703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187114.hg.1 |
TC1000012551.hg.1 |
Clariom D TC |