Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040116.hg.1 |
TC02002525.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040116.hg.1 |
ENST00000234198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040116.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040116.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040117.hg.1 |
TC02002525.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040117.hg.1 |
ENST00000234198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040117.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02040117.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014912.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209060.hg.1 |
TC0200014912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
ENST00000234198.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
ENST00000466293.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
NM_004405.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200209061.hg.1 |
TC0200014912.hg.1 |
Clariom D TC |