Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
TC0X001310.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
ENST00000320339.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
ENST00000371594.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
ENST00000476164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
ENST00000486230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
ENST00000536133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017589.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
TC0X001310.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
ENST00000371594.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
ENST00000536133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017590.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010133.hg.1 |
ENST00000521192.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172274.hg.1 |
TC0X00010640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010133.hg.1 |
ENST00000521192.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010133.hg.1 |
NR_003671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172272.hg.1 |
TC0X00010640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00172276.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172276.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172276.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172276.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172276.hg.1 |
TC0X00010640.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000320339.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000469448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
ENST00000536133.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170569.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_001170570.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010640.hg.1 |
NM_022101.3 |
Refseq |