Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073156.1 | 2 | 199 | 100 | ||
GenBank mRNA | AK125219.1 | - | 2 | 194 | 100 | |
AK296147.1 | - | 2 | 156 | 100 | ||
DA272550.1 | - | 2 | 128 | 100 | ||
DB248557.1 | - | 2 | 199 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |