Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
ENST00000317568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
ENST00000355991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
ENST00000416776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007797.hg.1 |
TC11000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007800.hg.1 |
ENST00000317568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007800.hg.1 |
ENST00000355991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007800.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007800.hg.1 |
TC11000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007801.hg.1 |
ENST00000317568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007801.hg.1 |
ENST00000355991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007801.hg.1 |
ENST00000416776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007801.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007801.hg.1 |
TC11000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
ENST00000317568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
ENST00000355991.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
ENST00000416776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007796.hg.1 |
TC11000627.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007799.hg.1 |
ENST00000317568.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007799.hg.1 |
ENST00000416776.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007799.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007799.hg.1 |
TC11000627.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000416776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000526201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000416776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000526201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157845.hg.1 |
TC1100008015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157838.hg.1 |
TC1100008015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000416776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000525156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000526201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000416776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000525156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000526201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000531296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_001300833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157847.hg.1 |
TC1100008015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000317568.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000355991.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000528854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
ENST00000533782.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
NM_001300832.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
NM_031904.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157852.hg.1 |
TC1100008015.hg.1 |
Clariom D TC |