Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017207.hg.1 |
ENST00000332601.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017207.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017207.hg.1 |
TC11001367.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017208.hg.1 |
ENST00000332601.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017208.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017208.hg.1 |
TC11001367.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175510.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175510.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175510.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175510.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175510.hg.1 |
TC1100009990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009990.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175511.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175511.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175511.hg.1 |
TC1100009990.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100175509.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175509.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175509.hg.1 |
ENST00000332601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175509.hg.1 |
NM_003696.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175509.hg.1 |
TC1100009990.hg.1 |
Clariom D TC |