Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020918.hg.1 |
ENST00000309403.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020918.hg.1 |
NM_001005469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020918.hg.1 |
TC11001807.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010941.hg.1 |
ENST00000309403.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010941.hg.1 |
NM_001005469.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010939.hg.1 |
ENST00000456493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182585.hg.1 |
ENST00000309403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182585.hg.1 |
NM_001005469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182585.hg.1 |
ENST00000309403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182585.hg.1 |
NM_001005469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182585.hg.1 |
TC1100010941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010941.hg.1 |
ENST00000309403.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010941.hg.1 |
NM_001005469.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010939.hg.1 |
ENST00000456493.1 |
Ensembl |