Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046301.hg.1 |
TC01002983.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046301.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046301.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213833.hg.1 |
TC0100015270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213834.hg.1 |
TC0100015270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000411951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
ENST00000436252.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015270.hg.1 |
NR_029429.1 |
Refseq |