Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008257.hg.1 |
TC02000527.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008257.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008258.hg.1 |
TC02000527.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008258.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222530.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222530.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222530.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222530.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222530.hg.1 |
TC0200016483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222531.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222531.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222531.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222531.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222531.hg.1 |
TC0200016483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
ENST00000616781.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016483.hg.1 |
NR_037931.1 |
Refseq |