Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
ENST00000375887.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
ENST00000430559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004129.hg.1 |
TC13000383.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
ENST00000542136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010000.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000375887.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000430559.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000479435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
ENST00000493765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
NM_001145645.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
NM_006573.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167072.hg.1 |
TC1300010000.hg.1 |
Clariom D TC |