Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030396.1 | 1 | 42 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 658 | MIR658 |
MIRN658, hsa-mir-658 |
-3.3kb | Sense | Chr. 22 - 37844272 - 37844371 on NCBI Build GRCh38 |
Downstream | eukaryotic translation initiation factor 3 subunit L | EIF3L |
EIF3EIP, EIF3S11, EIF3S6IP, HSPC021, HSPC025, MSTP005 |
674.0 | Anti-sense | Chr. 22 - 37848774 - 37888782 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |