Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7903403 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7903402 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7903401 |
- |
- |
Affymetrix Human Genome U219 Array |
11762639_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11762639_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014338.hg.1 |
TC01000913.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014338.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014339.hg.1 |
TC01000913.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014339.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009254.hg.1 |
ENST00000440278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165880.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165880.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165880.hg.1 |
TC0100009254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165882.hg.1 |
ENST00000440278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165882.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165882.hg.1 |
ENST00000440278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165882.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165882.hg.1 |
TC0100009254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100165883.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165883.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165883.hg.1 |
TC0100009254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009254.hg.1 |
ENST00000440278.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009254.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165881.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165881.hg.1 |
NR_033424.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165881.hg.1 |
TC0100009254.hg.1 |
Clariom D TC |