Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7903564 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7903563 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7903562 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7903561 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
231586_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014703.hg.1 |
TC01000935.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014703.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014703.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014699.hg.1 |
TC01000935.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014699.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014699.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014700.hg.1 |
TC01000935.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014700.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014700.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014700.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014701.hg.1 |
TC01000935.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014701.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014701.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014701.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166500.hg.1 |
TC0100009332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009332.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166498.hg.1 |
TC0100009332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100166501.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166501.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166501.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166501.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166501.hg.1 |
TC0100009332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100166502.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166502.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166502.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166502.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166502.hg.1 |
TC0100009332.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
ENST00000369989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
ENST00000417241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
NR_034168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
NR_049777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100166499.hg.1 |
TC0100009332.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
231586_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
231586_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231586_at3 |
- |
- |
Human Genome U95 Set |
52870_at |
- |
- |
Human Genome U95 Set |
52870_PM_at |
- |
- |
Human Genome U133 Set |
231586_at |
- |
- |
Human Genome U133 Set |
231586_PM_at |
- |
- |