Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000429132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000441379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000608043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000609694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
ENST00000610063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038311.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007396.hg.1 |
NR_038316.1 |
Refseq |