Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000805.4 | NP_000796.1 | 2 - 3 | 279 | 78 | |
GenBank mRNA | BC069724.1 | - | 2 - 3 | 244 | 7262518 | 78 |
BC069762.1 | - | 2 - 3 | 244 | 7262530 | 78 | |
BM768483.1 | - | 2 - 3 | 278 | 78 | ||
V00511.1 | - | 1 - 2 | 220 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |