Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010312.hg.1 |
ENST00000510421.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027323.hg.1 |
ENST00000356018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027323.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027323.hg.1 |
TC11002281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000456861.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012291.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194395.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194395.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194395.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194395.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194395.hg.1 |
TC1100012291.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010312.hg.1 |
ENST00000510421.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
ENST00000356018.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
ENST00000525791.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
NM_017589.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100194398.hg.1 |
TC1100012291.hg.1 |
Clariom D TC |