Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008462.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160238.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160238.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160238.hg.1 |
ENST00000477009.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160238.hg.1 |
NR_046230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160238.hg.1 |
TC0300008462.hg.1 |
Clariom D TC |