Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018196.hg.1 |
ENST00000260908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018196.hg.1 |
ENST00000361112.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018196.hg.1 |
ENST00000449046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018196.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018196.hg.1 |
TC17001372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018197.hg.1 |
ENST00000260908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018197.hg.1 |
ENST00000361112.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018197.hg.1 |
ENST00000449046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018197.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17018197.hg.1 |
TC17001372.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206060.hg.1 |
TC1700012387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012387.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000260908.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000587436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000590802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
ENST00000628453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
NM_001195790.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206056.hg.1 |
TC1700012387.hg.1 |
Clariom D TC |