Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024766.4 | NP_079042.1 | 5 - 6 | 592 | 109 | |
XM_011533111.2 | XP_011531413.1 | 5 - 6 | 558 | 109 | ||
XM_011533113.2 | XP_011531415.1 | 3 - 4 | 268 | 109 | ||
XM_017004982.1 | XP_016860471.1 | 3 - 4 | 2695 | 109 | ||
XM_017004983.1 | XP_016860472.1 | 4 - 5 | 807 | 109 | ||
GenBank mRNA | AK027104.1 | - | 5 - 6 | 532 | 109 | |
BC053733.1 | - | 5 - 6 | 519 | 5199179 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |