Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261827.1 | NP_001248756.1 | 3 - 4 | 980 | 96 | |
NM_001261828.1 | NP_001248757.1 | 4 - 5 | 1031 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |