Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012634.1 | NP_001012652.1 | 3 - 4 | 131 | 91 | |
NM_001012635.1 | NP_001012653.1 | 3 - 4 | 126 | 91 | ||
GenBank mRNA | AK296143.1 | - | 3 - 4 | 136 | 91 | |
JN546102.1 | - | 2 - 3 | 53 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |