Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156582.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156582.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156582.hg.1 |
TC1800008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156582.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156582.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156580.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156580.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156580.hg.1 |
TC1800008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156580.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156580.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156583.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156583.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156583.hg.1 |
TC1800008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156583.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156583.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156581.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156581.hg.1 |
NR_110796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156581.hg.1 |
TC1800008295.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156581.hg.1 |
ENST00000579713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156581.hg.1 |
NR_110796.1 |
Refseq |