Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
POP5 homolog, ribonuclease P/MRP subunit |
POP5 |
HSPC004, RPP2, RPP20, hPop5 |
-48.6kb | Sense | Chr. 12 - 120579045 - 120581452 on NCBI Build GRCh38 |
Downstream | calcium binding protein 1 | CABP1 |
CALBRAIN, HCALB_BR |
466.0 | Anti-sense | Chr. 12 - 120640552 - 120680869 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |