Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018603.hg.1 |
ENST00000331552.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018603.hg.1 |
ENST00000420605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018603.hg.1 |
ENST00000544883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018603.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018603.hg.1 |
TC19001301.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
TC1900010047.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900182828.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000331552.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000420605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000537399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000538165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000539693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
ENST00000544883.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_001300945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010047.hg.1 |
NM_176880.5 |
Refseq |