Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301410.1 | NP_001288339.1 | 2 - 3 | 409 | 148 | |
GenBank mRNA | AK301467.1 | - | 2 - 3 | 335 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |