Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04399399_m1
See other ARNTL GE Assays ›
Gene Symbol:
ARNTL
Gene Name:
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases:
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location:
Chr.11: 13276613 - 13387268 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04399399_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ARNTL
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001030272.2
NP_001025443.1
12 - 13
1469
193
NM_001030273.2
NP_001025444.1
13 - 14
1540
193
NM_001178.5
NP_001169.3
13 - 14
1524
193
NM_001297719.1
NP_001284648.1
13 - 14
1527
193
NM_001297722.1
NP_001284651.1
12 - 13
1472
193
NM_001297724.1
NP_001284653.1
13 - 14
1536
193
XM_006718233.3
XP_006718296.1
12 - 13
1414
193
XM_006718234.1
XP_006718297.1
13 - 14
1480
193
XM_011520105.2
XP_011518407.2
11 - 12
1575
193
XM_011520107.2
XP_011518409.2
11 - 12
1570
193
XM_011520108.1
XP_011518410.1
13 - 14
1520
193
XM_011520109.1
XP_011518411.1
11 - 12
1275
193
XM_011520110.1
XP_011518412.1
14 - 15
1535
193
XM_011520111.1
XP_011518413.1
12 - 13
1350
193
XM_011520112.2
XP_011518414.1
14 - 15
1535
193
XM_011520113.1
XP_011518415.1
14 - 15
1532
193
XM_017017738.1
XP_016873227.1
11 - 12
1576
193
XM_017017739.1
XP_016873228.1
11 - 12
1572
193
XM_017017740.1
XP_016873229.1
12 - 13
1411
193
XM_017017741.1
XP_016873230.1
11 - 12
1272
193
XM_017017742.1
XP_016873231.1
11 - 12
1275
193
XM_017017743.1
XP_016873232.1
11 - 12
1272
193
XM_017017744.1
XP_016873233.1
14 - 15
1532
193
XM_017017745.1
XP_016873234.1
13 - 14
1477
193
XM_017017746.1
XP_016873235.1
13 - 14
1490
193
XM_017017747.1
XP_016873236.1
12 - 13
1350
193
XM_017017748.1
XP_016873237.1
12 - 13
1404
193
GenBank mRNA
AB000812.1
-
10 - 11
1108
193
AF044288.1
-
13 - 14
1437
193
AK095749.1
-
13 - 14
1425
193
AK291510.1
-
12 - 13
1385
193
AK315174.1
-
13 - 14
1465
193
BC016674.1
-
10 - 11
1071
3921870
193
BC031214.1
-
13 - 14
1368
4793007
193
BC041129.1
-
13 - 14
1448
5753529
193
D89722.1
-
10 - 11
1084
193
U51627.1
-
7 - 8
1066
193
U60415.1
-
12 - 13
1371
193
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ARNTL
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
circadian rhythm
negative regulation of TOR signaling
circadian regulation of gene expression
regulation of protein catabolic process
regulation of hair cycle
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of neurogenesis
regulation of insulin secretion
regulation of cell cycle
response to redox state
maternal process involved in parturition
positive regulation of canonical Wnt signaling pathway
oxidative stress-induced premature senescence
regulation of type B pancreatic cell development
negative regulation of glucocorticoid receptor signaling pathway
regulation of cellular senescence
positive regulation of skeletal muscle cell differentiation
transcription regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
core promoter sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
protein binding
aryl hydrocarbon receptor binding
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
Hsp90 protein binding
repressing transcription factor binding
E-box binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0