Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297437.1 | NP_001284366.1 | 2 - 3 | 477 | 84 | |
NM_012176.2 | NP_036308.1 | 2 - 3 | 475 | 84 | ||
NM_033484.2 | NP_277019.1 | 2 - 3 | 475 | 84 | ||
XM_011514026.2 | XP_011512328.1 | 2 - 3 | 480 | 84 | ||
GenBank mRNA | AF176703.1 | - | 2 - 3 | 413 | 84 | |
AK024075.1 | - | 2 - 3 | 475 | 84 | ||
AU143782.1 | - | 2 - 3 | 475 | 84 | ||
BC040011.1 | - | 2 - 3 | 436 | 84 | ||
BC048098.1 | - | 2 - 3 | 443 | 4830192 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |