Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000440863.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000561725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000565881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000566479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000566631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000567472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000569437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000570276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001099436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001284364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001284365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000440863.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000561725.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000565881.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000566479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000566631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000567472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568667.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000568718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000569437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
ENST00000570276.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001099436.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001284364.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NM_001284365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104307.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104308.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
NR_104309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174811.hg.1 |
TC1500010045.hg.1 |
Clariom D TC |