Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000380302.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000406686.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000436008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000467144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000467177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000485307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000490273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000496753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000497691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_001286455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_015482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_021945.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NR_104448.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000380302.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000406686.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000436008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000467144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000467177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000485307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000490273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000496753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
ENST00000497691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_001286455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_015482.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NM_021945.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
NR_104448.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200378.hg.1 |
TC0600014223.hg.1 |
Clariom D TC |