Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000265344.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000373731.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000449081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000474988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000481911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000536244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_001286579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_001286580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_173562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NR_104480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NR_104481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000265344.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000373731.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000449081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000474988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000481911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
ENST00000536244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_001286579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_001286580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NM_173562.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NR_104480.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
NR_104481.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154372.hg.1 |
TC0600007836.hg.1 |
Clariom D TC |