Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102546294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_105057.1 | 3 - 4 | 656 | 70 | ||
GenBank mRNA | AK054753.1 | - | 3 - 4 | 589 | 70 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
serine peptidase inhibitor, Kazal type 13 (putative) |
SPINK13 |
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3 |
-4.0kb | Anti-sense | Chr. 5 - 148268860 - 148286210 on NCBI Build GRCh38 |
Downstream |
serine peptidase inhibitor, Kazal type 7 (putative) |
SPINK7 |
ECG2, ECRG2 |
4.3kb | Anti-sense | Chr. 5 - 148312427 - 148315919 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |