Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109834.1 | 1 | 1125 | 114 | ||
GenBank mRNA | GQ911592.1 | - | 1 | 1125 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |