Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008418.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160077.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160077.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160077.hg.1 |
ENST00000490351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160077.hg.1 |
NR_109998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160077.hg.1 |
TC0300008418.hg.1 |
Clariom D TC |