Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009630.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167364.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167364.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167364.hg.1 |
ENST00000449714.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167364.hg.1 |
NR_110240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167364.hg.1 |
TC0200009630.hg.1 |
Clariom D TC |