Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
ENST00000477616.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NM_004977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011194.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
NR_110912.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
TC1900011194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
ENST00000376959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
ENST00000474951.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193986.hg.1 |
NR_110912.1 |
Refseq |