Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010092.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175480.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175480.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175480.hg.1 |
ENST00000558424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500175480.hg.1 |
NR_120360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500175480.hg.1 |
TC1500010092.hg.1 |
Clariom D TC |