Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009018.hg.1 |
ENST00000443679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009018.hg.1 |
ENST00000443679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009018.hg.1 |
ENST00000443679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009018.hg.1 |
ENST00000443679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149144.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149144.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149144.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149144.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149144.hg.1 |
TC1300007245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
ENST00000603464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007245.hg.1 |
NR_120400.1 |
Refseq |