Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120622.1 | 2 - 3 | 297 | 107 | ||
GenBank mRNA | BE671563.1 | - | 1 - 2 | 107 | ||
BM794484.1 | - | 2 - 3 | 297 | 107 | ||
BX094877.1 | - | 2 - 3 | 274 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutathione S-transferase omega 2 | GSTO2 |
GSTO 2-2, bA127L20.1 |
-19.0kb | Sense | Chr. 10 - 104267626 - 104304948 on NCBI Build GRCh38 |
Downstream | cilia and flagella associated protein 58 | CFAP58 |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
11.8kb | Sense | Chr. 10 - 104339124 - 104455090 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons |