Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156936.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156936.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156936.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156936.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156936.hg.1 |
TC1300008446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156934.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156934.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156934.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156934.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156934.hg.1 |
TC1300008446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156935.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156935.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156935.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156935.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156935.hg.1 |
TC1300008446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
ENST00000499662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008446.hg.1 |
NR_047484.2 |
Refseq |