Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168183.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168183.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168183.hg.1 |
TC2000009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168183.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168183.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168182.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168182.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168182.hg.1 |
TC2000009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168182.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168182.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168185.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168185.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168185.hg.1 |
TC2000009776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168185.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168185.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
ENST00000431361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009776.hg.1 |
NR_102430.1 |
Refseq |