Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156421.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156421.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156421.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156421.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156421.hg.1 |
TC0400008206.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400156422.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156422.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156422.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156422.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156422.hg.1 |
TC0400008206.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400156423.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156423.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156423.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156423.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156423.hg.1 |
TC0400008206.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
ENST00000508933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008206.hg.1 |
NR_102713.1 |
Refseq |