Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179763.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179763.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179763.hg.1 |
TC0300010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300179766.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179766.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179766.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179766.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179766.hg.1 |
TC0300010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300179765.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179765.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179765.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179765.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179765.hg.1 |
TC0300010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300179767.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179767.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179767.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179767.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179767.hg.1 |
TC0300010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300179764.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179764.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179764.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179764.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179764.hg.1 |
TC0300010947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
ENST00000451485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010947.hg.1 |
NR_125406.1 |
Refseq |