Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006695.hg.1 |
ENST00000341449.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001650.hg.1 |
ENST00000341449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001650.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001650.hg.1 |
TC11000113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001649.hg.1 |
ENST00000341449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001649.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001649.hg.1 |
TC11000113.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100146744.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146744.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146744.hg.1 |
TC1100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146745.hg.1 |
ENST00000341449.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146745.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146745.hg.1 |
ENST00000341449.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146745.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146745.hg.1 |
TC1100006695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006695.hg.1 |
ENST00000341449.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006695.hg.1 |
NM_001004754.2 |
Refseq |