Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 1513 | 98 | |
GenBank mRNA | AJ566382.1 | - | 1 | 274 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |