Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000685.4 | NP_000676.1 | 2 - 3 | 339 | 79 | |
GenBank mRNA | AK291541.1 | - | 2 - 3 | 308 | 79 | |
BC068494.1 | - | 2 - 3 | 308 | 30337806 | 79 | |
DA649921.1 | - | 2 - 3 | 341 | 79 | ||
M93394.1 | - | 2 - 3 | 123 | 79 | ||
S77410.1 | - | 2 - 3 | 285 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |